Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1,3-Bis(2,6-diisopropylphenyl)-1H-imidazol-2(3H)-ylidene)silver(III) chloride
A588532-1G
A588532-250MG
77266-590EA
119.48
USD
77266-590
77266-592
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver 97%
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver
Precaución: for Research Use Only
Formula:
C₂₇H₃₆AgClN₂ MW: 531.92 g/mol |
MDL Number:
MFCD15072152 CAS Number: 873297-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...