(E)-2-((1-Benzylpiperidin-4-yl)methylene)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one 97%
Supplier: Ambeed
|
Warning
|
A588459-1G
A588459-250MG
A588459-5G
77266-584EA
87
USD
77266-584
77266-586
77266-588
(E)-2-((1-Benzylpiperidin-4-yl)methylene)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one 97%
(E)-2-((1-Benzylpiperidin-4-yl)methylene)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only
Formula:
C₂₄H₂₇NO₃ |
MDL Number:
MFCD08693144 CAS Number: 145546-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...