2-(2-Thienyl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Thiophen-2-yl)pyridine, 2-(2-Pyridyl)thiophene

A588357-10G A588357-1G A588357-250MG A588357-5G
77266-578EA 79.31 USD
77266-578 77266-580 77266-582 77749-207
2-(2-Thienyl)pyridine 97%
2-(2-Thienyl)pyridine

Precaución: for Research Use Only

Formula: C₉H₇NS
MW: 161.22 g/mol
MDL Number: MFCD00044441
CAS Number: 3319-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR