N-(3-((2-((4-(4-Methylpiperazin-1-yl)phenyl)amino)furo[3,2-d]pyrimidin-4-yl)oxy)phenyl)acrylamide 98+%

Supplier: Ambeed
Warning

Synonyms: Poseltinib

A587798-10MG A587798-1MG A587798-5MG
77266-562EA 71.07 USD
77266-562 77266-564 77266-566
N-(3-((2-((4-(4-Methylpiperazin-1-yl)phenyl)amino)furo[3,2-d]pyrimidin-4-yl)oxy)phenyl)acrylamide 98+%
N-(3-((2-((4-(4-Methylpiperazin-1-yl)phenyl)amino)furo[3,2-d]pyrimidin-4-yl)oxy)phenyl)acrylamide

Precaución: for Research Use Only

Formula: C₂₆H₂₆N₆O₃
MW: 470.52 g/mol
MDL Number: MFCD31729279
CAS Number: 1353552-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR