1-Butylpyridinium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butylpyridinium Chloride, N-Butylpyridinium chloride

A587556-100G A587556-1G A587556-25G A587556-5G
77242-476EA 69 USD
77242-476 77242-478 77242-480 77242-482
1-Butylpyridinium chloride 98%
1-Butylpyridinium chloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClN
MDL Number: MFCD00060047
CAS Number: 1124-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR