N-(5-Bromoimidazo[1,2-a]pyridin-2-yl)cyclopropanecarboxamide 97%

Supplier: Ambeed
Warning

A587457-100MG A587457-1G A587457-250MG
77266-556EA 54.59 USD
77266-556 77266-558 77266-560
N-(5-Bromoimidazo[1,2-a]pyridin-2-yl)cyclopropanecarboxamide 97%
N-(5-Bromoimidazo[1,2-a]pyridin-2-yl)cyclopropanecarboxamide

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrN₃O
MW: 280.12 g/mol
MDL Number: MFCD30531334
CAS Number: 1973485-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR