O411 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1

A587130-100MG A587130-10MG A587130-1MG A587130-25MG A587130-50MG A587130-5G
77242-466EA 84 USD
77242-466 77242-468 77242-470 77242-472 77242-474 77747-216
O411 98%
O411

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₂
MDL Number: MFCD00067778
CAS Number: 175135-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR