N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,4,6-triisopropylbenzenesulfonamide 97% +99%ee

Supplier: Ambeed
Warning

A586691-1G A586691-250MG A586691-5G
77242-456EA 37.08 USD
77242-456 77242-458 77242-460
N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,4,6-triisopropylbenzenesulfonamide 97% +99%ee
N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,4,6-triisopropylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₉H₃₈N₂O₂S
MW: 478.68 g/mol
MDL Number: MFCD22988918
CAS Number: 247923-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR