2-Amino-5-methylthiophene-3-carboxamide 97%

Supplier: Ambeed
Warning

A586086-100MG A586086-1G A586086-250MG A586086-5G
77254-160EA 6 USD
77254-160 77254-162 77254-164 77254-166
2-Amino-5-methylthiophene-3-carboxamide 97%
2-Amino-5-methylthiophene-3-carboxamide

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MDL Number: MFCD00082804
CAS Number: 51486-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR