3,5-Dimethoxybenzoyl chloride 98%

Supplier: Ambeed
Danger

A585835-25G A585835-5G A585835-1G
77280-182EA 109.18 USD
77280-182 77551-764 77749-205
3,5-Dimethoxybenzoyl chloride 98%
3,5-Dimethoxybenzoyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.62 g/mol
MDL Number: MFCD00000676
CAS Number: 17213-57-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR