4-(1,3-Oxazol-5-yl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(5-Oxazolyl)benzoic Acid

A585285-1G A585285-250MG A585285-25G A585285-5G
77292-660EA 39.14 USD
77292-660 77292-662 77292-664 77292-666
4-(1,3-Oxazol-5-yl)benzoic acid 98%
4-(1,3-Oxazol-5-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD02682033
CAS Number: 250161-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR