1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene 95%

Supplier: Ambeed
Warning

A585276-100MG A585276-250MG A585276-1G
77242-452EA 33 USD
77242-452 77242-454 77617-458
1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene 95%
1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
MDL Number: MFCD22551456
CAS Number: 1129541-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR