2-Bromoresorcinol 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromobenzene-1,3-diol

A584572-100G A584572-1G A584572-250MG A584572-5G A584572-25G
77266-544EA 509.85 USD
77266-544 77266-546 77266-548 77266-550 77551-758
2-Bromoresorcinol 96%
2-Bromoresorcinol

Precaución: for Research Use Only

Formula: C₆H₅BrO₂
MW: 189.008 g/mol
MDL Number: MFCD07779241
CAS Number: 6751-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR