4-Nitroisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A584474-100MG A584474-1G A584474-5G
77292-654EA 22 USD
77292-654 77292-656 77292-658
4-Nitroisoquinolin-1(2H)-one 97%
4-Nitroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₆N₂O₃
MDL Number: MFCD00030740
CAS Number: 55404-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR