2-(3-Bromo-4-methoxyphenyl)pyridine 96%

Supplier: Ambeed
Warning

A584318-1G A584318-250MG A584318-5G
77242-440EA 21.63 USD
77242-440 77242-442 77242-444
2-(3-Bromo-4-methoxyphenyl)pyridine 96%
2-(3-Bromo-4-methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.1200 g/mol
MDL Number: MFCD05864835
CAS Number: 168823-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR