4,4'-Dimethoxychalcone 95+%

Supplier: Ambeed
Danger

A583968-25G A583968-5G
77292-652EA 124 USD
77292-652 77749-202
4,4'-Dimethoxychalcone 95+%
4,4'-Dimethoxychalcone

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₃
MDL Number: MFCD00025815
CAS Number: 2373-89-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR