2,6-Dichloro-3-cyano-4-picoline 98%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine

A583825-1000G A583825-100G A583825-25G A583825-500G
77280-174EA 146.26 USD
77280-174 77280-176 77280-178 77280-180
2,6-Dichloro-3-cyano-4-picoline 98%
2,6-Dichloro-3-cyano-4-picoline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD00052633
CAS Number: 875-35-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR