3-Methylisonicotinamide 97%

Supplier: Ambeed
Warning

A583278-1G A583278-250MG
77280-170EA 82 USD
77280-170 77280-172
3-Methylisonicotinamide 97%
3-Methylisonicotinamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MDL Number: MFCD03411690
CAS Number: 251101-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR