1-Tetradecyne 98%

Supplier: Ambeed
Danger

A582732-1G A582732-25G A582732-5G
77242-434EA 22 USD
77242-434 77242-436 77242-438
1-Tetradecyne 98%
1-Tetradecyne

Precaución: for Research Use Only

Formula: C₁₄H₂₆
MDL Number: MFCD00015076
CAS Number: 765-10-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR