3-(Prop-2-ynyloxy)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Prop-2-yn-1-yloxy)pyridine

A582081-1G A582081-250MG
77242-430EA 33.99 USD
77242-430 77242-432
3-(Prop-2-ynyloxy)pyridine 97%
3-(Prop-2-ynyloxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.147 g/mol
MDL Number: MFCD10698154
CAS Number: 69022-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR