2-Bromo-3,5-difluorophenol 96%

Supplier: Ambeed
Warning

A581519-10G A581519-1G A581519-5G A581519-250MG
77266-536EA 152.44 USD
77266-536 77266-538 77266-540 77747-205
2-Bromo-3,5-difluorophenol 96%
2-Bromo-3,5-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
MDL Number: MFCD01861127
CAS Number: 325486-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR