R-(+)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin 98%

Supplier: Ambeed
Warning

Synonyms: (R)-SYNPHOS™

A581464-100MG A581464-250MG A581464-50MG
77266-530EA 109 USD
77266-530 77266-532 77266-534
R-(+)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin 98%
R-(+)-6,6'-Bis(diphenylphosphino)-2,2',3,3'-tetrahydro-5,5'-bi-1,4-benzodioxin

Precaución: for Research Use Only

Formula: C₄₀H₃₂O₄P₂
MW: 638.63 g/mol
MDL Number: MFCD05861608
CAS Number: 445467-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR