4,4'-Difluorobenzil 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-fluorophenyl)ethane-1,2-dione, Bis(4-fluorophenyl)ethanedione

A581204-100G A581204-10G A581204-1G A581204-25G A581204-5G
77292-642EA 380.07 USD
77292-642 77292-644 77292-646 77292-648 77594-768
4,4'-Difluorobenzil 98%
4,4'-Difluorobenzil

Precaución: for Research Use Only

Formula: C₁₄H₈F₂O₂
MW: 246.21 g/mol
MDL Number: MFCD00134541
CAS Number: 579-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR