7-Bromo-2-methylimidazo[1,2-a]pyridin-3-amine 97%

Supplier: Ambeed
Warning

A580489-100MG A580489-250MG
77242-426EA 136.99 USD
77242-426 77242-428
7-Bromo-2-methylimidazo[1,2-a]pyridin-3-amine 97%
7-Bromo-2-methylimidazo[1,2-a]pyridin-3-amine

Precaución: for Research Use Only

Formula: C₈H₈BrN₃
MW: 226.07 g/mol
MDL Number: MFCD22077072
CAS Number: 1504843-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR