2-Fluorobenzaldehyde 98%

Supplier: Ambeed
Danger

A580150-25G A580150-100G
77266-528EA 14 USD
77266-528 77594-760
2-Fluorobenzaldehyde 98%
2-Fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO
MW: 124.11 g/mol
Boiling Pt: 175
Melting Pt: -44.5
Flash Pt: 55
MDL Number: MFCD00003302
CAS Number: 446-52-6
UN: 1989
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR