KPT-8602 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(3-(3,5-Bis(trifluoromethyl)phenyl)-1H-1,2,4-triazol-1-yl)-2-(pyrimidin-5-yl)acrylamide, Eltanexor, (E)-3-[3-[3,5-bis(trifluoromethyl)phenyl]-1,2,4-triazol-1-yl]-2-pyrimidin-5-ylprop-2-enamide

A579961-10MG A579961-50MG A579961-5MG A579961-1MG A579961-100MG
77266-522EA 63.86 USD
77266-522 77266-524 77266-526 77617-444 77749-194
KPT-8602 98%
KPT-8602

Precaución: for Research Use Only

Formula: C₁₇H₁₀F₆N₆O
MW: 428.29 g/mol
MDL Number: MFCD30489739
CAS Number: 1642300-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR