TAK-063 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(2-fluoro-4-(1H-pyrazol-1-yl)phenyl)-5-methoxy-3-(1-phenyl-1H-pyrazol-5-yl)pyridazin-4(1H)-one, Balipodect

A579863-100MG A579863-10MG A579863-1MG A579863-25MG A579863-50MG A579863-5MG
77242-414EA 160.68 USD
77242-414 77242-416 77242-418 77242-420 77242-422 77242-424
TAK-063 98+%
TAK-063

Precaución: for Research Use Only

Formula: C₂₃H₁₇FN₆O₂
MW: 428.42 g/mol
MDL Number: MFCD28385852
CAS Number: 1238697-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR