4-Fluoro-3-toluoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-3-methylbenzoyl chloride, 4-Fluoro-3-methylbenzene-1-carbonyl chloride

A579816-1G A579816-5G A579816-25G
77292-634EA 18 USD
77292-634 77292-636 77594-756
4-Fluoro-3-toluoyl chloride 98%
4-Fluoro-3-toluoyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MDL Number: MFCD01631426
CAS Number: 455-84-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR