1,3-Dichloro-2-fluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 2,6-Dichlorofluorobenzene

A579645-25G A579645-5G
77254-138EA 8.24 USD
77254-138 77254-140
1,3-Dichloro-2-fluorobenzene 98%
1,3-Dichloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
MDL Number: MFCD00010307
CAS Number: 2268-05-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR