(Pivaloyloxy)methyl butyrate 95%

Supplier: Ambeed
Warning

Synonyms: Pivanex

A579458-1G A579458-250MG
77254-134EA 188.49 USD
77254-134 77254-136
(Pivaloyloxy)methyl butyrate 95%
(Pivaloyloxy)methyl butyrate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₄
MW: 202.24 g/mol
MDL Number: MFCD00209896
CAS Number: 122110-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR