Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 97%

Supplier: Ambeed
Warning

Synonyms: (2,6-Dichloro-5-fluoronicotinoyl)acetic Acid Ethyl Ester, Ethyl (2,6-Dichloro-5-fluoronicotinoyl)acetate, Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate, 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionic Acid Ethyl Ester

A579170-25G A579170-5G
77242-406EA 47.38 USD
77242-406 77242-408
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 97%
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂FNO₃
MW: 280.08 g/mol
MDL Number: MFCD00799535
CAS Number: 96568-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR