Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Nicardipine USP Related Compound A, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid Monomethyl ester, 5-(Methoxycarbonyl)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid

A578728-100G A578728-10G A578728-25G A578728-5G
77280-158EA 195.7 USD
77280-158 77280-160 77280-162 77749-188
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98%
Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₆
MW: 332.31 g/mol
MDL Number: MFCD00475478
CAS Number: 74936-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR