2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole 98+%

Supplier: Ambeed
Danger

A578663-100MG A578663-1G A578663-250MG A578663-5G
77242-386EA 17 USD
77242-386 77242-388 77242-390 77242-392
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole 98+%
2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole

Precaución: for Research Use Only

Formula: C₁₇H₂₆Cl₂O₂
MDL Number: MFCD03093939
CAS Number: 146370-52-7
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR