SC-236 97%

Supplier: Ambeed
Danger

Synonyms: 4-[5-(4-Chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulphonamide, 4-(5-(4-Chlorophenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide

A578005-10MG A578005-1MG A578005-50MG A578005-5MG
77242-360EA 30.9 USD
77242-360 77242-362 77242-364 77242-366
SC-236 97%
SC-236

Precaución: for Research Use Only

Formula: C₁₆H₁₁ClF₃N₃O₂S
MW: 401.8 g/mol
MDL Number: MFCD00941297
CAS Number: 170569-86-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR