4-Bromo-3-methylbenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-methyl-4-(trifluoromethyl)benzene

A577901-100G A577901-10G A577901-1G A577901-250MG A577901-25G A577901-5G
77242-348EA 773.53 USD
77242-348 77242-350 77242-352 77242-354 77242-356 77242-358
4-Bromo-3-methylbenzotrifluoride 97%
4-Bromo-3-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.0325 g/mol
MDL Number: MFCD09258773
CAS Number: 929000-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR