4-Ethyl-2,5-dimethoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A577749-100MG A577749-1G A577749-250MG A577749-5G
77266-502EA 34 USD
77266-502 77266-504 77266-506 77266-508
4-Ethyl-2,5-dimethoxybenzaldehyde 95%
4-Ethyl-2,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MDL Number: MFCD09743613
CAS Number: 50505-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR