1,1-Bis((2S,5S)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%

Supplier: Ambeed
Warning

A577275-100MG A577275-1G A577275-250MG
77254-112EA 143 USD
77254-112 77254-114 77254-116
1,1-Bis((2S,5S)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I) 98%
1,1-Bis((2S,5S)-2,5-diethylphospholano)ferrocene(cyclooctadiene)rhodium(I)

Precaución: for Research Use Only

Formula: C₃₄H₄₂FeP₂Rh
MW: 671.39 g/mol
MDL Number: MFCD17018833
CAS Number: 290347-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR