3-Methoxypyrrolidine 98%

Supplier: Ambeed
Danger

A576747-1G A576747-5G
77280-148EA 36 USD
77280-148 77749-184
3-Methoxypyrrolidine 98%
3-Methoxypyrrolidine

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD07787006
CAS Number: 62848-20-8
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR