5-Carboxy-2-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Borono-4-methoxybenzoic acid

A576565-1G A576565-250MG
77280-144EA 38 USD
77280-144 77280-146
5-Carboxy-2-methoxyphenylboronic acid 98%
5-Carboxy-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₅
MW: 195.9700 g/mol
MDL Number: MFCD06203261
CAS Number: 730971-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR