ML-277 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
ML277, (R)-N-(4-(4-methoxyphenyl)thiazol-2-yl)-1-tosylpiperidine-2-carboxamide, VU0458298-2, CID-53347902, (2R)-N-[4-(4-Methoxyphenyl)-1,3-thiazol-2-yl]-1-(4-methylphenyl)sulphonylpiperidine-2-carboxamide
A576326-1G
A576326-1MG
A576326-25MG
A576326-50MG
A576326-5MG
77242-316EA
1020.73
USD
77242-316
77242-318
77242-320
77242-322
77242-324
ML-277 98%
ML-277
Precaución: for Research Use Only
Formula:
C₂₃H₂₅N₃O₄S₂ MW: 471.59 g/mol |
MDL Number:
MFCD28133374 CAS Number: 1401242-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...