3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide
A575945-100MG
A575945-1G
A575945-250MG
77292-628EA
80.34
USD
77292-628
77292-630
77292-632
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] 98%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]
Precaución: for Research Use Only
Formula:
C₂₅H₂₇IN₂S₂ MW: 546.53 g/mol |
MDL Number:
MFCD00075033 CAS Number: 53213-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...