3,5-Dibromopyridine-N-oxide 98%

Supplier: Ambeed
Warning

A575903-5G A575903-1G A575903-25G
77280-142EA 70 USD
77280-142 77749-182 77749-183
3,5-Dibromopyridine-N-oxide 98%
3,5-Dibromopyridine N-oxide

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO
MDL Number: MFCD00160750
CAS Number: 2402-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR