(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methoxy-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-Me-Asp(OMe)-OH

A575752-1G A575752-250MG
77242-310EA 121 USD
77242-310 77242-312
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methoxy-4-oxobutanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methoxy-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₆
MDL Number: MFCD30475935
CAS Number: 2044710-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR