2-(Ethoxycarbonyl)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid
A575136-100G
A575136-10G
A575136-250MG
A575136-25G
A575136-5G
77254-102EA
298.7
USD
77254-102
77254-104
77254-106
77254-108
77254-110
2-(Ethoxycarbonyl)phenylboronic acid 98%
2-(Ethoxycarbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₁BO₄ MW: 193.99 g/mol |
MDL Number:
MFCD02179453 CAS Number: 380430-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...