3,4-Dichlorocinnamic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dichlorocinnamic Acid

A574837-100G A574837-10G A574837-25G A574837-5G
77266-486EA 136.99 USD
77266-486 77266-488 77266-490 77266-492
3,4-Dichlorocinnamic acid 97%
3,4-Dichlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
MDL Number: MFCD00004385
CAS Number: 1202-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR