Tetradecyl isobutyrate 97%

Supplier: Ambeed
Warning

A574579-100MG A574579-1G A574579-250MG
77242-288EA 13 USD
77242-288 77242-290 77242-292
Tetradecyl isobutyrate 97%
Tetradecyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₂
MW: 284.47 g/mol
MDL Number: MFCD30470137
CAS Number: 167871-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR