((2S,2'S)-1,1'-((2-Hydroxy-5-methyl-1,3-phenylene)bis(methylene))bis(pyrrolidine-2,1-diyl))bis(diphenylmethanol) 98%

Supplier: Ambeed
Warning

A574389-250MG A574389-100MG A574389-1G
77254-094EA 66.95 USD
77254-094 77617-428 77747-188
((2S,2'S)-1,1'-((2-Hydroxy-5-methyl-1,3-phenylene)bis(methylene))bis(pyrrolidine-2,1-diyl))bis(diphenylmethanol) 98%
((2S,2'S)-1,1'-((2-Hydroxy-5-methyl-1,3-phenylene)bis(methylene))bis(pyrrolidine-2,1-diyl))bis(diphenylmethanol)

Precaución: for Research Use Only

Formula: C₄₃H₄₆N₂O₃
MW: 638.84 g/mol
MDL Number: MFCD08276890
CAS Number: 320714-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR