2-Fluoro-1-methylpyridinium-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate

A574089-1G A574089-25G A574089-5G
77266-478EA 15.45 USD
77266-478 77266-480 77266-482
2-Fluoro-1-methylpyridinium-p-toluenesulfonate 98%
2-Fluoro-1-methylpyridinium p-toluenesulphonate

Precaución: for Research Use Only

Formula: C₁₃H₁₄FNO₃S
MW: 283.32 g/mol
MDL Number: MFCD00011983
CAS Number: 58086-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR