4-Butoxy-2,3-difluorophenol 97%

Supplier: Ambeed
Warning

A572348-100MG A572348-1G A572348-250MG
77242-264EA 63.86 USD
77242-264 77242-266 77242-268
4-Butoxy-2,3-difluorophenol 97%
4-Butoxy-2,3-difluorophenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.19 g/mol
MDL Number: MFCD07368023
CAS Number: 136239-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR