6-Benzyl-1-oxa-6-azaspiro[2.5]octane 98%

Supplier: Ambeed
Warning

A571969-1G A571969-250MG A571969-5G
77266-460EA 68 USD
77266-460 77266-462 77266-464
6-Benzyl-1-oxa-6-azaspiro[2.5]octane 98%
6-Benzyl-1-oxa-6-azaspiro[2.5]octane

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO
MDL Number: MFCD06808592
CAS Number: 19867-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR